β-Cyclodextrine, Sigma-Aldrich®
1
Catalog # SIALW402826-SAMPLE
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
- CAS Number:7585-39-9
- Formula:C₄₂H₇₀O₃₅
- MDL Number:MFCD00078139
- Molecular Weight:1134.98 g/mol
- :M-Clarity™ quality level = MQ300
- EINECS:231-493-2
Specifications
Specifications
- Pk:1



