Order Entry
Export
Orders LinkContactUsLinkComponent
Propionic acid, Sigma-Aldrich®

Propionic acid, Sigma-Aldrich®

Glass bottle for solids, 100 g
Catalog # SIALW292419-100G-K
Corrosive
Irritant
Supplier:   Merck
Propionic acid, Sigma-Aldrich®
Catalog # SIALW292419-100G-K
Supplier:  Merck
CAS Number:  79-09-4
Propionic acid, Sigma-Aldrich®

Glass bottle for solids, 100 g

CAS Number:  79-09-4
Price
98,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,07917
  • Boiling Pt:141 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:0.99 g/cm³ (20 °C)
  • Flash Pt:49 °C
  • UN:3463
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:201-176-3
  • REACH:01-2119486971-24
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications