Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-(-)-Sorbitol, Sigma-Aldrich®

Glass bottle for solids, 1 kg
Catalog # SIALW302902-1KG-K
Supplier:   Merck
D-(-)-Sorbitol, Sigma-Aldrich®
Catalog # SIALW302902-1KG-K
Supplier:  Merck
CAS Number:  50-70-4
D-(-)-Sorbitol, Sigma-Aldrich®
D-(-)-Sorbitol, Sigma-Aldrich®

Glass bottle for solids, 1 kg

CAS Number:  50-70-4
Price
93,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:94…98 °C
  • Density:1.524 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications