Order Entry
Export
Orders LinkContactUsLinkComponent
 

Acetone, Sigma-Aldrich®

Glass bottle, 100 g
Catalog # SIALW332615-100G
Flammable
Irritant
Supplier:   Merck
Acetone, Sigma-Aldrich®
Catalog # SIALW332615-100G
Supplier:  Merck
CAS Number:  67-64-1
Acetone, Sigma-Aldrich®
Acetone, Sigma-Aldrich®

Glass bottle, 100 g

CAS Number:  67-64-1
Price
100,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • UN:1090
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • REACH:01-2119471330-49
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications