Order Entry
Export
Orders LinkContactUsLinkComponent
Fumaric acid, Sigma-Aldrich®

Fumaric acid, Sigma-Aldrich®

25 kg
Catalog # SIALW248800-25KG-K
Irritant
Supplier:   Merck
Fumaric acid, Sigma-Aldrich®
Catalog # SIALW248800-25KG-K
Supplier:  Merck
CAS Number:  110-17-8
Fumaric acid, Sigma-Aldrich®

25 kg

CAS Number:  110-17-8
Price
726,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116.07 g/mol
  • Merck Index:13,04308
  • Boiling Pt:356 °C (1013 hPa)
  • Melting Pt:287 °C
  • Density:1.625 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  • Flash Pt:273 °C
  • EINECS:203-743-0
Specifications

Specifications

  • Pk:
    25 kg

Specifications