Order Entry
Export
Orders LinkContactUsLinkComponent
 

Caffeine, Sigma-Aldrich®

Glass bottle for solids, 1 kg
Catalog # SIALW222402-1KG-K
Irritant
Supplier:   Merck
Caffeine, Sigma-Aldrich®
Catalog # SIALW222402-1KG-K
Supplier:  Merck
CAS Number:  58-08-2
Caffeine, Sigma-Aldrich®
Caffeine, Sigma-Aldrich®

Glass bottle for solids, 1 kg

CAS Number:  58-08-2
Price
87,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  • CAS Number:58-08-2
  • Formula:C₈H₁₀N₄O₂
  • MDL Number:MFCD00005758
  • Molecular Weight:194.19 g/mol
  • Merck Index:13,01636
  • Melting Pt:235…239 °C
  • Density:1.23 g/cm³ (19 °C)
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • EINECS:200-362-1

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications