Order Entry
Export
Orders LinkContactUsLinkComponent
2,4,6-Trichlorophenol, Sigma-Aldrich®

2,4,6-Trichlorophenol, Sigma-Aldrich®

Glass bottle for solids, 250 g
Catalog # SIALT55301-250G
Health
Irritant
Environment
Supplier:   Merck
2,4,6-Trichlorophenol, Sigma-Aldrich®
Catalog # SIALT55301-250G
Supplier:  Merck
CAS Number:  88-06-2
2,4,6-Trichlorophenol, Sigma-Aldrich®

Glass bottle for solids, 250 g

CAS Number:  88-06-2
Price
96,00 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:88-06-2
  • Formula:C₆H₃Cl₃O
  • MDL Number:MFCD00002172
  • Molecular Weight:197.45 g/mol
  • Merck Index:12,09773
  • Boiling Pt:244.5 °C (1013 hPa)
  • Melting Pt:∼66 °C
  • Density:1.675 g/cm³ (25 °C)
  • Flash Pt:99 °C
  • Storage Temperature:Ambient
  • UN:2020
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:201-795-9

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications