Order Entry
Export
Orders LinkContactUsLinkComponent
Pentachlorophenol, Sigma-Aldrich®

Pentachlorophenol, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIALP2604-5G
Toxic
Health
Environment
Supplier:   Merck
Pentachlorophenol, Sigma-Aldrich®
Catalog # SIALP2604-5G
Supplier:  Merck
CAS Number:  87-86-5
Pentachlorophenol, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  87-86-5
Price
38,40 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:87-86-5
  • Formula:C₆HCl₅O
  • Molecular Weight:266.34 g/mol
  • Merck Index:12,07242
  • Boiling Pt:310 °C (1013 hPa)
  • Melting Pt:188 °C
  • Density:1.98 g/cm³ (20 °C)
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ200
  •  :Density: 1.98 g/cm³ (20 °C)
  • EINECS:201-778-6
  • UN:3155
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications