Order Entry
Export
Orders LinkContactUsLinkComponent
2-Nitro-m-xylene, Sigma-Aldrich®

2-Nitro-m-xylene, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIALN28353-5G
Supplier:   Merck
2-Nitro-m-xylene, Sigma-Aldrich®
Catalog # SIALN28353-5G
Supplier:  Merck
CAS Number:  81-20-9
2-Nitro-m-xylene, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  81-20-9
Price
33,90 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-Nitro-m-xylene, 1,3-Dimethyl-2-nitrobenzene, 2,6-Dimethylnitrobenzene
  • CAS Number:81-20-9
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00007181
  • Molecular Weight:151.16 g/mol
  • Boiling Pt:∼225 °C (1013 hPa)
  • Melting Pt:14…16 °C
  • Density:1.112 g/cm³ (20 °C)
  • Flash Pt:87 °C
  • Storage Temperature:Ambient
  • EINECS:201-333-6
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ100
  • UN:1665
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications