Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-Linalool, Sigma-Aldrich®

(±)-Linalool, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIALL2602-5G
Irritant
Supplier:   Merck
(±)-Linalool, Sigma-Aldrich®
Catalog # SIALL2602-5G
Supplier:  Merck
CAS Number:  78-70-6
(±)-Linalool, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  78-70-6
Price
52,20 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  • CAS Number:78-70-6
  • Formula:C₁₀H₁₈O
  • MDL Number:MFCD00008906
  • Molecular Weight:154.25 g/mol
  • Merck Index:13,05517
  • Boiling Pt:199 °C (1013 hPa)
  • Density:0.87 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Flash Pt:75 °C
  • Storage Temperature:Ambient
  • EINECS:201-134-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications