2'-Hydroxy-4'-methoxyacetophenone, Sigma-Aldrich®
Glass bottle for solids, 1 g
SIALH35803-1G
: Merck
: 1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
- :552-41-0
- :C₉H₁₀O₃
- :MFCD00008730
- :166.18 g/mol
- :154 °C (20 mmHg)
- :51 °C
- :1.1604 g/cm³ (40 °C)
- :M-Clarity™ quality level = MQ100
- :209-012-2
- Pack type:Glass bottle for solids
- Pk:1 g