Order Entry
Export
Orders LinkContactUsLinkComponent
 

2'-Hydroxy-4'-methoxyacetophenone, Sigma-Aldrich®

Glass bottle for solids, 10 g
Catalog # SIALH35803-10G
Supplier:   Merck
2'-Hydroxy-4'-methoxyacetophenone, Sigma-Aldrich®
Catalog # SIALH35803-10G
Supplier:  Merck
CAS Number:  552-41-0
2'-Hydroxy-4'-methoxyacetophenone, Sigma-Aldrich®

Glass bottle for solids, 10 g

CAS Number:  552-41-0
Price
96,10 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
  • CAS Number:552-41-0
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00008730
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:154 °C (20 mmHg)
  • Melting Pt:51 °C
  • Density:1.1604 g/cm³ (40 °C)
  •  :M-Clarity™ quality level = MQ100
  • EINECS:209-012-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications