Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Hydroxycyclohexanone dimer, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIALH24402-25G
Supplier:   Merck
2-Hydroxycyclohexanone dimer, Sigma-Aldrich®
Catalog # SIALH24402-25G
Supplier:  Merck
CAS Number:  30282-14-5
2-Hydroxycyclohexanone dimer, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  30282-14-5
Price
339,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:30282-14-5
  • Formula:C₆H₁₀O₂
  • MDL Number:MFCD00151157
  • Molecular Weight:228.28 g/mol
  • ADR:4.1,III
  •  :M-Clarity™ quality level = MQ100
  • UN:1325
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications