Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dichlorophenol, Sigma-Aldrich®

3,4-Dichlorophenol, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIALD70406-25G
Supplier:   Merck
3,4-Dichlorophenol, Sigma-Aldrich®
Catalog # SIALD70406-25G
Supplier:  Merck
CAS Number:  95-77-2
3,4-Dichlorophenol, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  95-77-2
Price
75,10 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:95-77-2
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002258
  • Molecular Weight:163 g/mol
  • Boiling Pt:247 °C (1013 hPa)
  • Melting Pt:66…68 °C
  • Density:1.6 g/cm³ (20 °C)
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ200
  •  :Density: 1.6 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-450-5
  • UN:2020
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications