Order Entry
Export
Orders LinkContactUsLinkComponent
2-Chlorocyclohexanone, Sigma-Aldrich®

2-Chlorocyclohexanone, Sigma-Aldrich®

Plastic bottle for solids, 100 g
Catalog # SIALC32607-100G
Supplier:   Merck
2-Chlorocyclohexanone, Sigma-Aldrich®
Catalog # SIALC32607-100G
Supplier:  Merck
CAS Number:  822-87-7
2-Chlorocyclohexanone, Sigma-Aldrich®

Plastic bottle for solids, 100 g

CAS Number:  822-87-7
Price
896,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:822-87-7
  • Formula:C₆H₉ClO
  • MDL Number:MFCD00001626
  • Molecular Weight:132.59 g/mol
  • Boiling Pt:203 °C (1013 hPa)
  • Melting Pt:18…20 °C
  • Density:1.16 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:212-505-5
  • UN:3335
  • ADR:9,
  •  :M-Clarity™ quality level = MQ100
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g

Specifications