Order Entry
Export
Orders LinkContactUsLinkComponent
4-Ethylpyrocatechol, Sigma-Aldrich®

4-Ethylpyrocatechol, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIAL683957-5G
Supplier:   Merck
4-Ethylpyrocatechol, Sigma-Aldrich®
Catalog # SIAL683957-5G
Supplier:  Merck
CAS Number:  1124-39-6
4-Ethylpyrocatechol, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  1124-39-6
Price
301,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
  • CAS Number:1124-39-6
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00015847
  • Molecular Weight:138.17 g/mol
  • EINECS:214-397-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications