Buffer, TRIS solution pH 7,6 (1 mol/L), Sigma-Aldrich®
TRIS buffer solution pH 7,6 (1 mol/l), Glass bottle, 100 mL
Catalog # SIALT2444-100ML
Supplier: Merck
Buffer, TRIS solution pH 7,6 (1 mol/L), Sigma-Aldrich®
Catalog # SIALT2444-100ML
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Description:TRIS buffer solution pH 7,6 (1 mol/l)
- Cat. No.:SIALT2444-100ML
- M-Clarity quality level:MQ200



