Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Indium tin oxide coated glass slide, 30-60 O/sq surface resistivity 1 * 25 items
Catalog # SIAL636908-25PAK
Supplier:
Merck
Expand image
Indium tin oxide coated glass slide, 30-60 O/sq surface resistivity 1 * 25 items
Catalog # SIAL636908-25PAK
Supplier:
Merck