ContactUsLinkComponent
RPMI-1640 MEDIUM, 10×, WITHOUT L-GLUTAMINE, FOLIC ACID AND SODIUM BICARBONATE, STERILE-FILTERED. CELL CULTURE TESTED 1 * 500 mL
Catalog # SIALR1145-500ML
Supplier:  Merck
RPMI-1640 MEDIUM, 10×, WITHOUT L-GLUTAMINE, FOLIC ACID AND SODIUM BICARBONATE, STERILE-FILTERED. CELL CULTURE TESTED 1 * 500 mL
Catalog # SIALR1145-500ML
Supplier:  Merck