Order Entry
Export
Orders LinkContactUsLinkComponent
Catechol violet indicator, Sigma-Aldrich®

Catechol violet indicator, Sigma-Aldrich®

Glass bottle for solids, 10 g
Catalog # P7884-10G
Irritant
Catechol violet indicator, Sigma-Aldrich®
Catalog # P7884-10G
CAS Number:  115-41-3
Catechol violet indicator, Sigma-Aldrich®
Catechol violet indicator, Sigma-Aldrich®

Glass bottle for solids, 10 g

CAS Number:  115-41-3
Price
178,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Pyrocatechol Violet (PV) is a sulfone phthalein dye. It chelates metal ions. The metal-PV complex is bluish-violet in colour.

  • Complexometric indicator
  • Pyrocatechol Violet has been used in the formation of dye-metal complexes
  • It has also been used to determine Cu²⁺ chelating activity
Synonyms:  Pyrocatechol Violet
  • CAS Number:115-41-3
  • Formula:C₁₉H₁₄O₇S
  • MDL Number:MFCD00005868
  • Molecular Weight:386.38 g/mol
  • Melting Pt:185 °C (dec., lit.)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Room Temperature
  • EINECS:204-088-3
Specifications

Specification Test Results

  • Appearance (Form):
    Powder
  • 1H NMR Spectrum:
    Conforms to Structure
  • Solubility (Colour), c = 10 mg/10 ml; Water:
    Faint Brown to Brown and Faint Red to Dark Red
  • Suitability, Suitable for use as an indicator for EDTA titrations:
    Suitable
  • Suitability, Suitable for use as a pH indicator:
    Suitable

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications