Order Entry
Export
Orders LinkContactUsLinkComponent
Melatonin, Sigma-Aldrich®

Melatonin, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIALM5250-5G
Supplier:   Merck
Melatonin, Sigma-Aldrich®
Catalog # SIALM5250-5G
Supplier:  Merck
CAS Number:  73-31-4
Melatonin, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  73-31-4
Price
551,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS Number:73-31-4
  • Formula:C₁₃H₁₆N₂O₂
  • MDL Number:MFCD00005655
  • Molecular Weight:232.28 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:200-797-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications