Melatonin, Sigma-Aldrich®
Glass bottle for solids, 10 g
SIALM5250-10G
: Merck
Jump to:
Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
- CAS Number:73-31-4
- Formula:C₁₃H₁₆N₂O₂
- MDL Number:MFCD00005655
- Molecular Weight:232.28 g/mol
- :M-Clarity™ quality level = MQ200
- EINECS:200-797-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



