Order Entry
Export
Orders LinkContactUsLinkComponent
1,2-Ethanedithiol, Sigma-Aldrich®

1,2-Ethanedithiol, Sigma-Aldrich®

Glass bottle, 100 mL
Catalog # SIAL398020-100ML
Flammable
Toxic
Supplier:   Merck
1,2-Ethanedithiol, Sigma-Aldrich®
Catalog # SIAL398020-100ML
Supplier:  Merck
CAS Number:  540-63-6
1,2-Ethanedithiol, Sigma-Aldrich®

Glass bottle, 100 mL

CAS Number:  540-63-6
Price
68,60 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  1.2-Ethanedithiol, 1,2-Dimercaptoethane
  • CAS Number:540-63-6
  • Formula:C₂H₆S₂
  • MDL Number:MFCD00004892
  • Molecular Weight:94.2 g/mol
  • Merck Index:13,03760
  • Boiling Pt:146…149 °C (1013 hPa)
  • Melting Pt:–41 °C
  • Density:1.374 g/cm³ (20 °C)
  • Flash Pt:44 °C
  • Storage Temperature:Ambient
  • UN:3071
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ200
  • EINECS:208-752-3
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications