Order Entry
Export
Orders LinkContactUsLinkComponent
(2-Methoxyethoxy)methyl chloride, Sigma-Aldrich®

(2-Methoxyethoxy)methyl chloride, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIAL357480-25G
Supplier:   Merck
(2-Methoxyethoxy)methyl chloride, Sigma-Aldrich®
Catalog # SIAL357480-25G
Supplier:  Merck
CAS Number:  3970-21-6
(2-Methoxyethoxy)methyl chloride, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  3970-21-6
Price
183,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1-Chloromethoxy-2-methoxyethane, 1-Chloro-2,5-dioxahexane, MEM chloride
  • CAS Number:3970-21-6
  • Formula:ClCH₂OCH₂CH₂OCH₃
  • MDL Number:MFCD00000888
  • Molecular Weight:124.57 g/mol
  • Boiling Pt:50 °C (1013 hPa)
  • Density:1.094 g/cm³ (20 °C)
  • EINECS:223-589-8
  • UN:1992
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ200
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications