Hydroxyurea, Sigma-Aldrich®
Glass bottle for solids, 25 g
SIALH8627-25G
: Merck
Jump to:
Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76.05 g/mol
- Merck Index:13,04874
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:142…144 °C
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:204-821-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



