Hydroxyurea, Sigma-Aldrich®
Glass bottle for solids, 10 g
SIALH8627-10G
: Merck
: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- :127-07-1
- :CH₄N₂O₂
- :MFCD00007943
- :76.05 g/mol
- :13,04874
- :296 °C (1013 hPa)
- :142…144 °C
- :M-Clarity™ quality level = MQ200
- :Ambient
- :204-821-7
- Pack type:Glass bottle for solids
- Pk:10 g