Order Entry
Export
Orders LinkContactUsLinkComponent
Hydroxyurea, Sigma-Aldrich®

Hydroxyurea, Sigma-Aldrich®

Glass bottle for solids, 100 g
Catalog # SIALH8627-100G
Supplier:   Merck
Hydroxyurea, Sigma-Aldrich®
Catalog # SIALH8627-100G
Supplier:  Merck
CAS Number:  127-07-1
Hydroxyurea, Sigma-Aldrich®

Glass bottle for solids, 100 g

CAS Number:  127-07-1
Price
1 610,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:13,04874
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:142…144 °C
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications