Ectoine, Sigma-Aldrich®
Glass bottle for solids, 100 mg
Catalog # SIALE2271-100MG
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
- CAS Number:96702-03-3
- Formula:C₆H₁₀N₂O₂
- MDL Number:MFCD03419286
- Molecular Weight:142.1558 g/mol
- :M-Clarity™ quality level = MQ200
- EINECS:431-910-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



