Order Entry
Export
Orders LinkContactUsLinkComponent
Digitonin, Sigma-Aldrich®

Digitonin, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIALD5628-5G
Supplier:   Merck
Digitonin, Sigma-Aldrich®
Catalog # SIALD5628-5G
Supplier:  Merck
CAS Number:  11024-24-1
Digitonin, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  11024-24-1
Price
1 940,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Digitonin
  • CAS Number:11024-24-1
  • Formula:C₅₆H₉₂O₂₉
  • MDL Number:MFCD00077729
  • Molecular Weight:1229.312 g/mol
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ200
  • EINECS:234-255-6
  • UN:3462
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications