D-(-)-Mannitol, Sigma-Aldrich®
Glass bottle for solids, 1 kg
Catalog # SIALM9647-1KG
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:13,05769
- Boiling Pt:290…295 °C (4 hPa)
- Melting Pt:164…169 °C
- Density:1.49 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:200-711-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



