Order Entry
Export
Orders LinkContactUsLinkComponent
Pyrocatechol, Sigma-Aldrich®

Pyrocatechol, Sigma-Aldrich®

Glass bottle for solids, 1 kg
Catalog # SIALC9510-1KG
Irritant
Supplier:   Merck
Pyrocatechol, Sigma-Aldrich®
Catalog # SIALC9510-1KG
Supplier:  Merck
CAS Number:  120-80-9
Pyrocatechol, Sigma-Aldrich®

Glass bottle for solids, 1 kg

CAS Number:  120-80-9
Price
268,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,08089
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:103…105 °C
  • Density:1.0548 g/cm³ (20 °C)
  • Flash Pt:127 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:204-427-5

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications