Order Entry
Export
Orders LinkContactUsLinkComponent
β-Cyclodextrine, Sigma-Aldrich®

β-Cyclodextrine, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIALC4805-5G
Supplier:   Merck
β-Cyclodextrine, Sigma-Aldrich®
Catalog # SIALC4805-5G
Supplier:  Merck
CAS Number:  7585-39-9
β-Cyclodextrine, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  7585-39-9
Price
92,60 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:231-493-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications