Order Entry
Export
Orders LinkContactUsLinkComponent
Sebacoyl dichloride, Sigma-Aldrich®

Sebacoyl dichloride, Sigma-Aldrich®

Glass bottle for solids, 500 g
Catalog # SIAL131784-500G
Corrosive
Irritant
Supplier:   Merck
Sebacoyl dichloride, Sigma-Aldrich®
Catalog # SIAL131784-500G
Supplier:  Merck
CAS Number:  111-19-3
Sebacoyl dichloride, Sigma-Aldrich®

Glass bottle for solids, 500 g

CAS Number:  111-19-3
Price
336,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Sebacoyl chloride
  • CAS Number:111-19-3
  • Formula:C₁₀H₁₆Cl₂O₂
  • MDL Number:MFCD00000770
  • Molecular Weight:239.14 g/mol
  • Boiling Pt:220 °C (1013 hPa)
  • Melting Pt:–5 °C
  • Density:1.12 g/cm³ (20 °C)
  • Flash Pt:165 °C
  • Storage Temperature:Ambient
  • EINECS:203-843-4
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ200
  • UN:3265
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications