Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cycloheximide, Sigma-Aldrich®

Glass bottle for solids, 1 g
Catalog # SIALC1988-1G
Toxic
Health
Environment
Supplier:   Merck
Cycloheximide, Sigma-Aldrich®
Catalog # SIALC1988-1G
Supplier:  Merck
CAS Number:  66-81-9
Cycloheximide, Sigma-Aldrich®
Cycloheximide, Sigma-Aldrich®

Glass bottle for solids, 1 g

CAS Number:  66-81-9
Price
118,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Ambient
  • ADR:6.1,I
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-636-0
  • UN:2811
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications