Order Entry
Export
Orders LinkContactUsLinkComponent
 

Selenium trioxide, Sigma-Aldrich®

Glass bottle for solids, 100 g
Catalog # SIAL213365-100G
Toxic
Health
Environment
Supplier:   Merck
Selenium trioxide, Sigma-Aldrich®
Catalog # SIAL213365-100G
Supplier:  Merck
CAS Number:  13768-86-0
Selenium trioxide, Sigma-Aldrich®

Glass bottle for solids, 100 g

CAS Number:  13768-86-0
Price
95,60 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Selenium dioxide
  • CAS Number:13768-86-0
  • Formula:O₃Se
  • Molecular Weight:126.96 g/mol
  • Melting Pt:117.85…118.85 °C
  • Density:3.6 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  •  :Density: 3.6 g/cm³ (20 °C)
  • EINECS:237-385-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications