Orders LinkContactUsLinkComponent
Glass beads
Glass beads, 5 mm
  SIAL18406-6X500G
 :   Merck
Glass beads
  SIAL18406-6X500G
 :  Merck
Glass beads
Glass beads, 5 mm
308,00 €
1 * 3.000 g
Made of PYREX® glass, solid.
For boiling and distillation operations. Use as distillation packing, mixing beads or boiling stones.
  • Ø:
    5 mm
  • Description:
    Glass beads
  • Seq:
    02