Order Entry
Export
Orders LinkContactUsLinkComponent
Iron(III) bromide, Sigma-Aldrich®

Iron(III) bromide, Sigma-Aldrich®

Glass bottle for solids, 50 g
Catalog # SIAL217883-50G
Irritant
Supplier:   Merck
Iron(III) bromide, Sigma-Aldrich®
Catalog # SIAL217883-50G
Supplier:  Merck
CAS Number:  10031-26-2
Iron(III) bromide, Sigma-Aldrich®

Glass bottle for solids, 50 g

CAS Number:  10031-26-2
Price
397,00 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Ferric bromide, Iron tribromide
  • CAS Number:10031-26-2
  • Formula:Br₃Fe
  • MDL Number:MFCD00016082
  • Molecular Weight:295.56 g/mol
  • Merck Index:12,04060
  • ADR:8,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:233-089-1
  • UN:3260
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications