Order Entry
Export
Orders LinkContactUsLinkComponent
Agar, Sigma-Aldrich®

Agar, Sigma-Aldrich®

Glass bottle for solids, 500 g
Catalog # SIALA9799-500G
Supplier:   Merck
Agar, Sigma-Aldrich®
Catalog # SIALA9799-500G
Supplier:  Merck
CAS Number:  9002-18-0
Agar, Sigma-Aldrich®
Agar, Sigma-Aldrich®

Glass bottle for solids, 500 g

CAS Number:  9002-18-0
Price
367,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • Formula:C₁₂nH₁₈nOn₉n
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications