Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) oxide, Sigma-Aldrich®

Lead(II) oxide, Sigma-Aldrich®

Glass bottle for solids, 10 g
Catalog # SIAL203610-10G
Health
Irritant
Environment
Supplier:   Merck
Lead(II) oxide, Sigma-Aldrich®
Catalog # SIAL203610-10G
Supplier:  Merck
CAS Number:  1317-36-8
Lead(II) oxide, Sigma-Aldrich®

Glass bottle for solids, 10 g

CAS Number:  1317-36-8
Price
98,50 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Lead monoxide
  • CAS Number:1317-36-8
  • Formula:PbO
  • MDL Number:MFCD00011164
  • Molecular Weight:223.2 g/mol
  • Merck Index:13,05431
  • Boiling Pt:1470 °C (1013 hPa)
  • Melting Pt:888 °C
  • Density:9.5 g/cm³ (20 °C)
  • EINECS:215-267-0
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
  • UN:2291
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications