Order Entry
Export
Orders LinkContactUsLinkComponent
Bronopol, Sigma-Aldrich®

Bronopol, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIAL134708-25G
Corrosive
Irritant
Environment
Supplier:   Merck
Bronopol, Sigma-Aldrich®
Catalog # SIAL134708-25G
Supplier:  Merck
CAS Number:  52-51-7
Bronopol, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  52-51-7
Price
36,50 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol
  • CAS Number:52-51-7
  • Formula:HOCH₂BrC(NO₂)CH₂OH
  • MDL Number:MFCD00007390
  • Molecular Weight:199.99 g/mol
  • Merck Index:13,01432
  • Boiling Pt:388 °C (1013 hPa)
  • Melting Pt:130…133 °C
  • Density:1.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-143-0
  • ADR:4.1,III
  •  :M-Clarity™ quality level = MQ200
  • UN:3241

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications