Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dichlorophenol, Sigma-Aldrich®

2,4-Dichlorophenol, Sigma-Aldrich®

Glass bottle for solids, 250 g
Catalog # SIAL105953-250G
Toxic
Corrosive
Environment
Supplier:   Merck
2,4-Dichlorophenol, Sigma-Aldrich®
Catalog # SIAL105953-250G
Supplier:  Merck
CAS Number:  120-83-2
2,4-Dichlorophenol, Sigma-Aldrich®

Glass bottle for solids, 250 g

CAS Number:  120-83-2
Price
82,70 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene
  • CAS Number:120-83-2
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002169
  • Molecular Weight:163 g/mol
  • Merck Index:13,03098
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:40…41 °C
  • Density:1.4 g/cm³ (20 °C)
  • Flash Pt:114 °C
  • Storage Temperature:Ambient
  • UN:2928
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ200
  • EINECS:204-429-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications