2,4-Dichlorophenol, Sigma-Aldrich®
Glass bottle for solids, 100 g
SIAL105953-100G
: Merck
: 1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene
- :120-83-2
- :C₆H₄Cl₂O
- :MFCD00002169
- :163 g/mol
- :13,03098
- :210 °C (1013 hPa)
- :40…41 °C
- :1.4 g/cm³ (20 °C)
- :114 °C
- :Ambient
- :2928
- :6.1,II
- :M-Clarity™ quality level = MQ200
- :204-429-6
- Pack type:Glass bottle for solids
- Pk:100 g