pH buffers, ready to use
Calibration and maintenance set with technical buffer solution (SORT/TPL/TRACE)
3 bottles with each 250 ml: pH 4,01/7,00/10,01 Trace,
1 bottle with 250 ml KCl solution 3 mol/L,
1 bottle with 250 ml pepsin cleaning solution, Plastic dosing bottle, 1 SET
3 bottles with each 250 ml: pH 4,01/7,00/10,01 Trace,
1 bottle with 250 ml KCl solution 3 mol/L,
1 bottle with 250 ml pepsin cleaning solution, Plastic dosing bottle, 1 SET
Catalog # WTWA108824
Supplier: WTW
Jump to:
Product Details & Documents
Every bottle has its own lot reference with the expiration date.
- Shelf life up to 3 years (unopened)
- Reference temperature at 25 °C
- Economic consumption
- Easiest handling by use of the dosing vessel
- One litre container with a separate compartment for the sample
Accuracy is ±0,03 pH
: Traceable to NIST.
Specifications
Specifications
- Pack type:Plastic dosing bottle
- Pk:1 SET
- Description:Calibration and maintenance set with technical buffer solution (SORT/TPL/TRACE)<br />3 bottles with each 250 ml: pH 4,01/7,00/10,01 Trace,<br />1 bottle with 250 ml KCl solution 3 mol/L,<br />1 bottle with 250 ml pepsin cleaning solution



