Polysorbate 80, J.T.Baker®
Glass bottle, amber, narrow mouth, 4 L
Supplier: Avantor
Jump to:
Product Details & Documents
Polyoxyethylene (20) Sorbitan Monooleate
Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Storage Temperature:Ambient
- EINECS:500-018-3
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Identification (by IR):Passes Test
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L



