Arrow 38202 ESD Occupational Clogs
49, White
Catalog # ABEB38202/49
Supplier: Abeba
Jump to:
Product Details & Documents
ESD clogs made from smooth leather with microfiber lining, microfibre heel patch and a foldable and adjustable heel strap.
- Non-slip PU/PU walking sole
- Sanitised treated
- Respiration absorbing
ESD dissipative according to DIN EN 61340.
Certifications: CE, EN ISO 20347:2012, OB, A,FO, SRA.
Documents
38202
Datasheet
Specifications
- Shoe size:49
- Colour:White
- Seq:02