Arrow 38202 ESD Occupational Clogs
48, White
ABEB38202/48
: Abeba
ESD clogs made from smooth leather with microfiber lining, microfibre heel patch and a foldable and adjustable heel strap.
- Non-slip PU/PU walking sole
- Sanitised treated
- Respiration absorbing
ESD dissipative according to DIN EN 61340.
: CE, EN ISO 20347:2012, OB, A,FO, SRA.
- Shoe size:48
- Colour:White
- Seq:01