Order Entry
Export
Orders LinkContactUsLinkComponent
Photographic trays

Photographic trays

250×200×60 mm
Catalog # DELT19281
Supplier:   DELTALAB
Photographic trays
Catalog # DELT19281
Supplier:  DELTALAB
Photographic trays

250×200×60 mm

Price
49,60 €
Log in for availability
1 * 1 items
Product Details & Documents

Photographic trays made of PVC.

  • Temperature resistance from −20 to +80 °C

With ribbed interior surface.

Specifications
  • Seq:
    02
  • W×D×H:
    250×200×60 mm