Orders LinkContactUsLinkComponent
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
14/23 NS, 14/23 NS, 110 mm
Catalog # 201-0597
Supplier:   SciLabware
Adapters, receiver, vacuum, Quickfit®
Catalog # 201-0597
Supplier:  SciLabware
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
14/23 NS, 14/23 NS, 110 mm
Price
73,60 €
Log in for availability
1 * 1 items
Product Details & Documents
PYREX®, borosilicate glass, bent.
  • Easy connection to flexible tubing via detachable plastic screw thread connector
Adapter with a side arm for connection to a vacuum supply.
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    14/23 NS
  • Total length:
    110 mm