Spherical ground joints, Rotulex®
O-ring PTFE, for joint size 19, Spherical joints RM:RF19
Catalog # BIBBRO19
Supplier: SciLabware
Jump to:
Product Details & Documents
Made of borosilicate glass.
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200 °C
- System comprises male ball joint with PTFE covered 'O' ring which fits into a female cup joint
- Joint can be retained together with spring loaded metal clip
Rotulex® offers a grease free spherical joint.
Ordering information: Component parts should be ordered separately .
Specifications
- For:Spherical joints RM:RF19
- Type:for joint size 19
- Description:O-ring PTFE
- Seq:17