Tween® 20 (Polysorbate), Sigma-Aldrich®
Glass bottle for solids, 1 kg
Catalog # EXPO93773
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Tween 20 MB Grade 1092800100 1 * 100 mL Catalog #655204-100
Product 655204-100 is similar to EXPO93773 but manufacturer and specifications may vary.
TWEEN 20 FOR SYNTHESIS 1 * 1 L Catalog #8.22184.1000
Product 8.22184.1000 is similar to EXPO93773 but manufacturer and specifications may vary.
Tween 20 (Polysorbate) EMPROVE ESSENTIAL Ph Eur,JPE,NF 1 * 2,5 L Catalog #8.17072.2500
See more alternativesProduct 8.17072.2500 is similar to EXPO93773 but manufacturer and specifications may vary.
Product Details & Documents
Synonyms: Polyoxyethylene sorbitan monolaurate
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Molecular Weight:1227.53 g/mol
- Density:1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:500-018-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



